Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
R3HDM2
Entrez Gene ID
22864
Gene Name
R3H domain containing 2
Gene Aliases
CAG6, PR01365
Chromosome Location
Chr.12: 57253764 - 57430999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011538031.1 XP_011536333.1 3 - 4 297 70
XM_011538033.1 XP_011536335.1 3 - 4 297 70
XM_011538035.1 XP_011536337.1 3 - 4 297 70
XM_011538036.1 XP_011536338.1 3 - 4 297 70
XM_011538038.1 XP_011536340.1 3 - 4 297 70
XM_011538039.1 XP_011536341.1 3 - 4 297 70
XM_011538040.1 XP_011536342.1 3 - 4 297 70
XM_011538042.2 XP_011536344.1 4 - 5 393 70
XM_017018999.1 XP_016874488.1 4 - 5 434 70
XM_017019000.1 XP_016874489.1 3 - 4 297 70
XM_017019003.1 XP_016874492.1 3 - 4 297 70
XM_017019004.1 XP_016874493.1 3 - 4 297 70
XM_017019008.1 XP_016874497.1 3 - 4 297 70
XM_017019009.1 XP_016874498.1 3 - 4 297 70
XM_017019011.1 XP_016874500.1 5 - 6 450 70
XM_017019012.1 XP_016874501.1 3 - 4 297 70
XM_017019019.1 XP_016874508.1 4 - 5 391 70
XM_017019020.1 XP_016874509.1 3 - 4 297 70
XM_017019026.1 XP_016874515.1 4 - 5 376 70

View Full Product Details