The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024763.4 | NP_079039.4 | 11 - 12 | 1785 | 79 |
XM_005271212.3 | XP_005271269.1 | 10 - 11 | 1543 | 79 | |
XM_011542161.2 | XP_011540463.1 | 10 - 11 | 1651 | 79 | |
XM_011542162.2 | XP_011540464.1 | 11 - 12 | 1738 | 79 | |
XM_017002353.1 | XP_016857842.1 | 10 - 11 | 1543 | 79 | |
XM_017002354.1 | XP_016857843.1 | 9 - 10 | 1456 | 79 | |
XM_017002355.1 | XP_016857844.1 | 8 - 9 | 1158 | 79 | |
XM_017002356.1 | XP_016857845.1 | 9 - 10 | 1402 | 79 | |
GenBank mRNA | AK026782.1 | - | 7 - 8 | 1026 | 79 |
AK292830.1 | - | 12 - 13 | 1738 | 79 | |
AL133617.1 | - | 2 - 3 | 181 | 79 | |
BC126379.1 | - | 11 - 12 | 1760 | 79 | |
BC126381.1 | - | 11 - 12 | 1760 | 79 | |
BX648840.1 | - | 11 - 12 | 1785 | 79 |