The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024763.4 | NP_079039.4 | 2 - 3 | 394 | 137 |
NM_207014.2 | NP_996897.2 | 2 - 3 | 394 | 137 | |
XM_005271212.3 | XP_005271269.1 | 2 - 3 | 347 | 137 | |
XM_011542161.2 | XP_011540463.1 | 2 - 3 | 347 | 137 | |
XM_011542162.2 | XP_011540464.1 | 2 - 3 | 347 | 137 | |
XM_017002353.1 | XP_016857842.1 | 2 - 3 | 347 | 137 | |
XM_017002354.1 | XP_016857843.1 | 2 - 3 | 347 | 137 | |
XM_017002356.1 | XP_016857845.1 | 1 - 2 | 168 | 137 | |
XM_017002357.1 | XP_016857846.1 | 2 - 3 | 347 | 137 | |
GenBank mRNA | AK127011.1 | - | 1 - 2 | 96 | 137 |
AK292830.1 | - | 3 - 4 | 347 | 137 | |
AL516734.3 | - | 2 - 3 | 341 | 137 | |
BC032406.1 | - | 2 - 3 | 293 | 137 | |
BC126379.1 | - | 2 - 3 | 369 | 137 | |
BC126381.1 | - | 2 - 3 | 369 | 137 | |
BX648840.1 | - | 2 - 3 | 395 | 137 |