The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167911.1 | NP_001161383.1 | 3 - 4 | 579 | 116 |
NM_001167912.1 | NP_001161384.1 | 3 - 4 | 579 | 116 | |
NM_001167915.1 | NP_001161387.1 | 4 - 5 | 926 | 116 | |
NM_001167916.1 | NP_001161388.1 | 3 - 4 | 664 | 116 | |
NM_001167917.1 | NP_001161389.1 | 2 - 3 | 508 | 116 | |
NM_024621.2 | NP_078897.2 | 3 - 4 | 770 | 116 | |
XM_011513134.2 | XP_011511436.1 | 2 - 3 | 513 | 116 | |
XM_011513135.1 | XP_011511437.1 | 3 - 4 | 409 | 116 | |
GenBank mRNA | AB051479.1 | - | 3 - 4 | 579 | 116 |
AK027625.1 | - | 3 - 4 | 668 | 116 | |
AL713656.1 | - | 2 - 3 | 367 | 116 | |
BC057999.1 | - | 3 - 4 | 667 | 116 | |
BC101660.1 | - | 2 - 3 | 406 | 116 | |
BC111017.1 | - | 3 - 4 | 571 | 116 | |
BC113555.1 | - | 2 - 3 | 406 | 116 |