The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004447.5 | NP_004438.3 | 17 - 18 | 2251 | 64 |
XM_011520605.2 | XP_011518907.1 | 18 - 19 | 2311 | 64 | |
GenBank mRNA | AK291777.1 | - | 17 - 18 | 2056 | 64 |
AK292931.1 | - | 17 - 18 | 1908 | 64 | |
AK301834.1 | - | 10 - 11 | 1241 | 64 | |
AK304860.1 | - | 17 - 18 | 2251 | 64 | |
AK316134.1 | - | 10 - 11 | 1160 | 64 | |
AK316239.1 | - | 10 - 11 | 1433 | 64 | |
BC030010.2 | - | 17 - 18 | 2063 | 64 | |
U12535.1 | - | 17 - 18 | 2023 | 64 |