Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBBP8
Entrez Gene ID
5932
Gene Name
RB binding protein 8, endonuclease
Gene Aliases
COM1, CTIP, JWDS, RIM, SAE2, SCKL2
Chromosome Location
Chr.18: 22914121 - 23026486 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002894.2 NP_002885.1 10 - 11 1244 118
NM_203291.1 NP_976036.1 10 - 11 1214 118
NM_203292.1 NP_976037.1 10 - 11 1295 118
XM_005258325.2 XP_005258382.1 10 - 11 1175 118
XM_005258326.3 XP_005258383.1 9 - 10 1105 118
XM_006722519.2 XP_006722582.1 12 - 13 1316 118
XM_006722520.2 XP_006722583.1 11 - 12 1252 118
XM_006722521.2 XP_006722584.1 10 - 11 1312 118
XM_011526132.2 XP_011524434.1 11 - 12 1263 118
XM_017025915.1 XP_016881404.1 8 - 9 794 118
XM_017025916.1 XP_016881405.1 11 - 12 1335 118
GenBank mRNA AF043431.1 - 10 - 11 1211 118
AK292481.1 - 10 - 11 1175 118
BC001170.1 - 10 - 11 1400 118
BC030590.1 - 10 - 11 1042 118
BX648221.1 - 10 - 11 1269 118
U72066.1 - 10 - 11 1212 118

View Full Product Details