Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHCR24
Entrez Gene ID
1718
Gene Name
24-dehydrocholesterol reductase
Gene Aliases
DCE, Nbla03646, SELADIN1, seladin-1
Chromosome Location
Chr.1: 54849627 - 54887248 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014762.3 NP_055577.1 5 - 6 1008 92
GenBank mRNA AB073391.1 - 6 - 7 883 92
AF261758.1 - 5 - 6 978 92
AK302774.1 - 5 - 6 839 92
AK304437.1 - 5 - 6 725 92
AK316427.1 - 5 - 6 709 92
BC004375.1 - 5 - 6 914 92
BC011669.2 - 5 - 6 941 92
D13643.3 - 5 - 6 991 92

View Full Product Details