Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMD5
Entrez Gene ID
5711
Gene Name
proteasome 26S subunit, non-ATPase 5
Gene Aliases
S5B
Chromosome Location
Chr.9: 120798318 - 120843021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005047.3 NP_005038.1 4 - 5 671 124
XM_011518866.2 XP_011517168.1 4 - 5 581 124
XM_017014927.1 XP_016870416.1 4 - 5 581 124
GenBank mRNA AK291051.1 - 4 - 5 572 124
BC014478.1 - 4 - 5 560 124
D31889.1 - 4 - 5 558 124
S79862.1 - 4 - 5 562 124

View Full Product Details