Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMD5
Entrez Gene ID
5711
Gene Name
proteasome 26S subunit, non-ATPase 5
Gene Aliases
S5B
Chromosome Location
Chr.9: 120798318 - 120843021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270427.1 NP_001257356.1 7 - 8 1102 99
NM_005047.3 NP_005038.1 8 - 9 1231 99
XM_011518866.2 XP_011517168.1 8 - 9 1141 99
XM_017014927.1 XP_016870416.1 8 - 9 1141 99
XM_017014928.1 XP_016870417.1 7 - 8 1031 99
GenBank mRNA AK026925.1 - 4 - 5 534 99
AK291051.1 - 8 - 9 1132 99
AK303007.1 - 7 - 8 1032 99
BC014478.1 - 8 - 9 1120 99
D31889.1 - 8 - 9 1118 99
S79862.1 - 8 - 9 1122 99

View Full Product Details