The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000352.4 | NP_000343.2 | 6 - 7 | 1082 | 73 |
NM_001287174.1 | NP_001274103.1 | 6 - 7 | 1082 | 73 | |
XM_011520331.2 | XP_011518633.1 | 6 - 7 | 1104 | 73 | |
XM_017018197.1 | XP_016873686.1 | 6 - 7 | 1076 | 73 | |
XM_017018198.1 | XP_016873687.1 | 6 - 7 | 1076 | 73 | |
XM_017018199.1 | XP_016873688.1 | 6 - 7 | 1081 | 73 | |
XM_017018200.1 | XP_016873689.1 | 6 - 7 | 1100 | 73 | |
XM_017018201.1 | XP_016873690.1 | 6 - 7 | 1076 | 73 | |
XM_017018202.1 | XP_016873691.1 | 5 - 6 | 863 | 73 | |
XM_017018203.1 | XP_016873692.1 | 6 - 7 | 1076 | 73 | |
GenBank mRNA | AB209084.1 | - | 6 - 7 | 1056 | 73 |
AF087138.1 | - | 6 - 7 | 1048 | 73 | |
AK297560.1 | - | 6 - 7 | 1041 | 73 | |
L78207.1 | - | 6 - 7 | 1051 | 73 | |
U63421.1 | - | 6 - 7 | 1013 | 73 |