The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030649.2 | NP_085152.2 | 19 - 20 | 1882 | 75 |
XM_005244717.3 | XP_005244774.1 | 7 - 8 | 876 | 75 | |
XM_011540606.2 | XP_011538908.1 | 19 - 20 | 1931 | 75 | |
XM_011540607.1 | XP_011538909.1 | 19 - 20 | 4355 | 75 | |
XM_011540608.2 | XP_011538910.1 | 18 - 19 | 1836 | 75 | |
XM_017000233.1 | XP_016855722.1 | 19 - 20 | 2071 | 75 | |
GenBank mRNA | BC004874.2 | - | 4 - 5 | 486 | 75 |
BC021851.1 | - | 2 - 3 | 272 | 75 | |
BC047001.1 | - | 9 - 10 | 1334 | 75 | |
BC051194.1 | - | 19 - 20 | 1882 | 75 |