Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR3
Entrez Gene ID
10885
Gene Name
WD repeat domain 3
Gene Aliases
DIP2, UTP12
Chromosome Location
Chr.1: 117929749 - 117960426 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006784.2 NP_006775.1 11 - 12 1398 123
GenBank mRNA AF083217.1 - 11 - 12 1379 123
AK022858.1 - 3 - 4 403 123
AK292438.1 - 11 - 12 1393 123

View Full Product Details