The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006784.2 | NP_006775.1 | 26 - 27 | 2737 | 94 |
GenBank mRNA | AB209201.1 | - | 14 - 15 | 1242 | 94 |
AF083217.1 | - | 26 - 27 | 2718 | 94 | |
AK022858.1 | - | 18 - 19 | 1742 | 94 | |
AK292438.1 | - | 26 - 27 | 2732 | 94 |