The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006784.2 | NP_006775.1 | 6 - 7 | 748 | 124 |
GenBank mRNA | AF083217.1 | - | 6 - 7 | 729 | 124 |
AK292438.1 | - | 6 - 7 | 743 | 124 | |
BC058836.1 | - | 7 - 8 | 787 | 124 |