Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NADSYN1
Entrez Gene ID
55191
Gene Name
NAD synthetase 1
Gene Aliases
-
Chromosome Location
Chr.11: 71453171 - 71501538 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018161.4 NP_060631.2 11 - 12 1128 96
GenBank mRNA AB091316.1 - 11 - 12 1002 96
AK001493.1 - 11 - 12 1069 96
AK022436.1 - 11 - 12 1098 96
AK094022.1 - 2 - 3 1110 96
AK097946.1 - 3 - 4 1007 96
BC003638.2 - 11 - 12 1091 96
BC003666.2 - 11 - 12 1091 96
CR457286.1 - 11 - 12 1002 96

View Full Product Details