Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NADSYN1
Entrez Gene ID
55191
Gene Name
NAD synthetase 1
Gene Aliases
-
Chromosome Location
Chr.11: 71453171 - 71501538 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018161.4 NP_060631.2 13 - 14 1281 91
GenBank mRNA AB091316.1 - 13 - 14 1155 91
AK001493.1 - 13 - 14 1222 91
AK022436.1 - 13 - 14 1251 91
AK094022.1 - 4 - 5 1263 91
AK097946.1 - 5 - 6 1160 91
AL512694.1 - 1 - 2 1220 91
BC003638.2 - 13 - 14 1244 91
BC003666.2 - 13 - 14 1244 91
CR457286.1 - 13 - 14 1155 91

View Full Product Details