The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018161.4 | NP_060631.2 | 9 - 10 | 925 | 72 |
GenBank mRNA | AB091316.1 | - | 9 - 10 | 799 | 72 |
AK001493.1 | - | 9 - 10 | 866 | 72 | |
AK022436.1 | - | 9 - 10 | 895 | 72 | |
BC003638.2 | - | 9 - 10 | 888 | 72 | |
BC003666.2 | - | 9 - 10 | 888 | 72 | |
CR457286.1 | - | 9 - 10 | 799 | 72 |