The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017977.2 | NP_001017977.1 | 1 - 2 | 457 | 118 |
NM_001198956.1 | NP_001185885.1 | 1 - 2 | 457 | 118 | |
NM_001198957.1 | NP_001185886.1 | 1 - 2 | 457 | 118 | |
NM_018442.3 | NP_060912.2 | 1 - 2 | 457 | 118 | |
XM_005245331.4 | XP_005245388.1 | 1 - 2 | 398 | 118 | |
XM_005245332.4 | XP_005245389.1 | 1 - 2 | 398 | 118 | |
XM_005245333.4 | XP_005245390.1 | 1 - 2 | 398 | 118 | |
GenBank mRNA | AF116725.1 | - | 1 - 2 | 198 | 118 |
AK093970.1 | - | 1 - 2 | 343 | 118 | |
AK226181.1 | - | 1 - 2 | 308 | 118 | |
AK297847.1 | - | 1 - 2 | 317 | 118 | |
AY766164.1 | - | 1 - 2 | 197 | 118 | |
DA128283.1 | - | 1 - 2 | 457 | 118 | |
DQ768089.1 | - | 1 - 2 | 121 | 118 |