Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTN1A1
Entrez Gene ID
696
Gene Name
butyrophilin subfamily 1 member A1
Gene Aliases
BT, BTN, BTN1
Chromosome Location
Chr.6: 26499290 - 26510425 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001732.2 NP_001723.2 3 - 4 728 99
XM_005249340.3 XP_005249397.2 4 - 5 1834 99
GenBank mRNA BC096312.3 - 3 - 4 728 99
BC096313.1 - 3 - 4 728 99
BC096314.1 - 3 - 4 728 99
BC096315.1 - 3 - 4 728 99
U39576.1 - 4 - 5 780 99

View Full Product Details