The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020533.2 | NP_065394.1 | 6 - 7 | 914 | 62 |
GenBank mRNA | AF249319.1 | - | 6 - 7 | 857 | 62 |
AF287269.1 | - | 6 - 7 | 898 | 62 | |
AJ293659.1 | - | 5 - 6 | 1088 | 62 | |
AJ293970.1 | - | 6 - 7 | 853 | 62 | |
AK026102.1 | - | 6 - 7 | 868 | 62 | |
AK222673.1 | - | 6 - 7 | 893 | 62 | |
AK294330.1 | - | 6 - 7 | 879 | 62 | |
BC005149.2 | - | 6 - 7 | 896 | 62 |