The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012455.2 | NP_036587.2 | 6 - 7 | 2017 | 66 |
XM_005263634.2 | XP_005263691.1 | 6 - 7 | 2082 | 66 | |
XM_006712392.2 | XP_006712455.1 | 6 - 7 | 2029 | 66 | |
XM_006712393.2 | XP_006712456.1 | 6 - 7 | 2029 | 66 | |
XM_006712394.2 | XP_006712457.1 | 6 - 7 | 2029 | 66 | |
GenBank mRNA | AB208798.1 | - | 5 - 6 | 2456 | 66 |
AJ459781.1 | - | 5 - 6 | 1855 | 66 | |
AK023421.1 | - | 4 - 5 | 906 | 66 | |
AK290045.1 | - | 6 - 7 | 2027 | 66 | |
BC035307.1 | - | 6 - 7 | 1919 | 66 | |
BC073151.1 | - | 6 - 7 | 1979 | 66 | |
U63127.1 | - | 5 - 6 | 1897 | 66 |