Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPECC1L
Entrez Gene ID
23384
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1 like
Gene Aliases
CYTSA, GBBB2, OBLFC1
Chromosome Location
Chr.22: 24270817 - 24417740 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145468.3 NP_001138940.3 14 - 15 3407 80
NM_001254733.1 NP_001241662.1 7 - 8 946 80
NM_015330.4 NP_056145.4 15 - 16 3511 80
GenBank mRNA AB002374.1 - 11 - 12 2736 80
AK025531.1 - 11 - 12 1815 80
AL832425.1 - 15 - 16 3430 80
AY884293.1 - 14 - 15 3335 80
BC136471.1 - 14 - 15 3277 80

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
Chromosome Location
Chr.22: 24270817 - 24442360 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_103546.1 16 - 17 3633 80
GenBank mRNA AB209022.1 - 16 - 17 3552 80

View Full Product Details