The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287437.1 | NP_001274366.1 | 5 - 6 | 733 | 72 |
NM_001287439.1 | NP_001274368.1 | 5 - 6 | 887 | 72 | |
NM_001287440.1 | NP_001274369.1 | 5 - 6 | 684 | 72 | |
NM_001287441.1 | NP_001274370.1 | 5 - 6 | 648 | 72 | |
NM_001287442.1 | NP_001274371.1 | 5 - 6 | 641 | 72 | |
NM_001287443.1 | NP_001274372.1 | 5 - 6 | 645 | 72 | |
NM_024900.4 | NP_079176.2 | 5 - 6 | 769 | 72 | |
NM_199320.3 | NP_955352.1 | 5 - 6 | 769 | 72 | |
XM_005263232.1 | XP_005263289.1 | 5 - 6 | 611 | 72 | |
XM_017008626.1 | XP_016864115.1 | 5 - 6 | 726 | 72 | |
XM_017008627.1 | XP_016864116.1 | 5 - 6 | 773 | 72 | |
GenBank mRNA | AB058710.1 | - | 1 - 2 | 69 | 72 |
AF520952.1 | - | 5 - 6 | 666 | 72 | |
AK027620.1 | - | 5 - 6 | 689 | 72 | |
AK127326.1 | - | 5 - 6 | 541 | 72 | |
AK304235.1 | - | 5 - 6 | 602 | 72 | |
AK304815.1 | - | 5 - 6 | 558 | 72 | |
BC032376.1 | - | 5 - 6 | 603 | 72 | |
DB293142.1 | - | 5 - 6 | 623 | 72 |