Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE1
Entrez Gene ID
79960
Gene Name
jade family PHD finger 1
Gene Aliases
PHF17
Chromosome Location
Chr.4: 128809623 - 128875224 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287437.1 NP_001274366.1 10 - 11 1873 139
NM_001287439.1 NP_001274368.1 10 - 11 2027 139
NM_001287440.1 NP_001274369.1 10 - 11 1824 139
NM_001287442.1 NP_001274371.1 10 - 11 1781 139
NM_001287443.1 NP_001274372.1 10 - 11 1785 139
NM_199320.3 NP_955352.1 10 - 11 1909 139
XM_005263232.1 XP_005263289.1 10 - 11 1751 139
GenBank mRNA AB058710.1 - 6 - 7 1209 139
AK127326.1 - 10 - 11 1681 139
AK304235.1 - 10 - 11 1742 139
AK304815.1 - 10 - 11 1698 139

View Full Product Details