Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE1
Entrez Gene ID
79960
Gene Name
jade family PHD finger 1
Gene Aliases
PHF17
Chromosome Location
Chr.4: 128809623 - 128875224 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287442.1 NP_001274371.1 1 - 2 132 64
GenBank mRNA DA714593.1 - 1 - 2 132 64
DB293142.1 - 1 - 2 114 64

View Full Product Details