Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NINL
Entrez Gene ID
22981
Gene Name
ninein like
Gene Aliases
NLP
Chromosome Location
Chr.20: 25452697 - 25585531 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318226.1 NP_001305155.1 17 - 18 2386 90
NM_025176.5 NP_079452.3 18 - 19 3433 90
XM_011529186.2 XP_011527488.1 19 - 20 4272 90
XM_011529187.2 XP_011527489.1 19 - 20 4257 90
XM_011529188.1 XP_011527490.1 19 - 20 3649 90
XM_011529189.1 XP_011527491.1 19 - 20 3657 90
XM_011529190.1 XP_011527492.1 17 - 18 3371 90
XM_011529191.1 XP_011527493.1 15 - 16 3164 90
XM_011529192.1 XP_011527494.1 12 - 13 4084 90
XM_011529193.1 XP_011527495.1 16 - 17 2324 90
XM_011529194.2 XP_011527496.1 10 - 11 3353 90
XM_011529195.2 XP_011527497.1 9 - 10 2178 90
XM_011529197.2 XP_011527499.1 11 - 12 2547 90
XM_011529198.1 XP_011527500.1 5 - 6 1719 90
XM_017027732.1 XP_016883221.1 20 - 21 5591 90
XM_017027733.1 XP_016883222.1 20 - 21 3703 90
XM_017027734.1 XP_016883223.1 12 - 13 2537 90
XM_017027735.1 XP_016883224.1 9 - 10 2183 90
GenBank mRNA AB023197.1 - 18 - 19 3419 90
BC036380.2 - 17 - 18 3365 90
EU718622.1 - 17 - 18 2372 90

View Full Product Details