The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020762.2 | NP_065813.1 | 2 - 3 | 322 | 104 |
XM_005269042.3 | XP_005269099.1 | 2 - 3 | 711 | 104 | |
XM_011538580.1 | XP_011536882.1 | 3 - 4 | 430 | 104 | |
XM_011538581.2 | XP_011536883.1 | 3 - 4 | 1935 | 104 | |
GenBank mRNA | AB037725.1 | - | 1 - 2 | 1613 | 104 |
BC029919.1 | - | 2 - 3 | 280 | 104 | |
BC053903.1 | - | 2 - 3 | 322 | 104 |