The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167582.1 | NP_001161054.1 | 4 - 5 | 434 | 84 |
NM_005777.2 | NP_005768.1 | 8 - 9 | 1947 | 84 | |
XM_005264784.1 | XP_005264841.1 | 8 - 9 | 2222 | 84 | |
XM_005264785.1 | XP_005264842.1 | 7 - 8 | 638 | 84 | |
XM_005264786.1 | XP_005264843.1 | 6 - 7 | 536 | 84 | |
XM_005264787.2 | XP_005264844.1 | 5 - 6 | 1302 | 84 | |
XM_006712916.1 | XP_006712979.1 | 8 - 9 | 2047 | 84 | |
XM_017005496.1 | XP_016860985.1 | 6 - 7 | 1376 | 84 | |
XM_017005497.1 | XP_016860986.1 | 4 - 5 | 271 | 84 | |
XM_017005498.1 | XP_016860987.1 | 5 - 6 | 460 | 84 | |
XM_017005499.1 | XP_016860988.1 | 10 - 11 | 2507 | 84 | |
XM_017005500.1 | XP_016860989.1 | 9 - 10 | 2389 | 84 | |
XM_017005501.1 | XP_016860990.1 | 9 - 10 | 2340 | 84 | |
XM_017005502.1 | XP_016860991.1 | 4 - 5 | 335 | 84 | |
GenBank mRNA | AF042857.1 | - | 8 - 9 | 1800 | 84 |
AF069517.1 | - | 8 - 9 | 1903 | 84 | |
AF091264.1 | - | 8 - 9 | 1832 | 84 | |
AK093836.1 | - | 6 - 7 | 479 | 84 | |
AK291719.1 | - | 8 - 9 | 1947 | 84 | |
AK298108.1 | - | 3 - 4 | 340 | 84 | |
AK303371.1 | - | 7 - 8 | 1766 | 84 | |
AK308794.1 | - | 7 - 8 | 562 | 84 | |
AL832236.1 | - | 4 - 5 | 383 | 84 | |
BC046643.1 | - | 4 - 5 | 288 | 84 | |
BX648535.1 | - | 8 - 9 | 2124 | 84 | |
DA068167.1 | - | 4 - 5 | 326 | 84 | |
U50839.1 | - | 7 - 8 | 1733 | 84 |