The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133369.2 | NP_588610.2 | 13 - 14 | 2382 | 143 |
XM_006714928.1 | XP_006714991.1 | 14 - 15 | 2530 | 143 | |
GenBank mRNA | BC009333.2 | - | 7 - 8 | 1288 | 143 |
BC122556.1 | - | 7 - 8 | 1259 | 143 | |
BC157824.1 | - | 13 - 14 | 2265 | 143 |