The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276414.1 | NP_001263343.1 | 5 - 6 | 1135 | 108 |
XM_005255505.1 | XP_005255562.1 | 14 - 15 | 2110 | 108 | |
XM_011522610.1 | XP_011520912.1 | 15 - 16 | 2170 | 108 | |
XM_011522612.1 | XP_011520914.1 | 14 - 15 | 1938 | 108 | |
XM_017023568.1 | XP_016879057.1 | 5 - 6 | 936 | 108 | |
XM_017023569.1 | XP_016879058.1 | 5 - 6 | 1051 | 108 | |
GenBank mRNA | AK022685.1 | - | 3 - 4 | 670 | 108 |
AK124977.1 | - | 4 - 5 | 1215 | 108 | |
BC000150.1 | - | 5 - 6 | 1097 | 108 |