The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016544.2 | NP_057628.1 | 5 - 6 | 712 | 84 |
XM_017004269.1 | XP_016859758.1 | 5 - 6 | 743 | 84 | |
XM_017004270.1 | XP_016859759.1 | 5 - 6 | 603 | 84 | |
GenBank mRNA | AF178983.1 | - | 5 - 6 | 546 | 84 |
AK294726.1 | - | 5 - 6 | 496 | 84 | |
BC034049.1 | - | 5 - 6 | 573 | 84 |