Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSBN1
Entrez Gene ID
54665
Gene Name
round spermatid basic protein 1
Gene Aliases
ROSBIN
Chromosome Location
Chr.1: 113761832 - 113812476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018364.4 NP_060834.2 6 - 7 1999 144
NR_130896.1 7 - 8 2117 144
GenBank mRNA AK002082.1 - 6 - 7 1814 144
AK022166.1 - 5 - 6 1178 144
AK292552.1 - 6 - 7 1999 144
BC026155.1 - 7 - 8 1939 144
CR627402.1 - 7 - 8 1592 144

View Full Product Details