The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025098.3 | NP_079374.2 | 3 - 4 | 542 | 90 |
XM_011545267.1 | XP_011543569.1 | 3 - 4 | 542 | 90 | |
GenBank mRNA | AK128620.1 | - | 2 - 3 | 999 | 90 |
AK291998.1 | - | 3 - 4 | 518 | 90 | |
AY157608.1 | - | 3 - 4 | 472 | 90 | |
BC103876.1 | - | 3 - 4 | 472 | 90 | |
BC103877.1 | - | 3 - 4 | 472 | 90 | |
BC103878.1 | - | 3 - 4 | 472 | 90 |