The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015429.3 | NP_056244.2 | 24 - 25 | 2114 | 118 |
XM_005247290.2 | XP_005247347.1 | 56 - 57 | 4196 | 118 | |
XM_005247301.2 | XP_005247358.1 | 54 - 55 | 4070 | 118 | |
XM_006713569.2 | XP_006713632.1 | 55 - 56 | 4118 | 118 | |
XM_011512657.1 | XP_011510959.1 | 53 - 54 | 3971 | 118 | |
XM_017006107.1 | XP_016861596.1 | 54 - 55 | 4064 | 118 | |
GenBank mRNA | AK299938.1 | - | 34 - 35 | 2196 | 118 |
BC030221.1 | - | 24 - 25 | 2094 | 118 |