The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286563.1 | NP_001273492.1 | 17 - 18 | 2426 | 57 |
NM_001286564.1 | NP_001273493.1 | 16 - 17 | 2135 | 57 | |
NM_020716.2 | NP_065767.1 | 16 - 17 | 2405 | 57 | |
XM_011542933.1 | XP_011541235.1 | 14 - 15 | 2547 | 57 | |
XM_017018038.1 | XP_016873527.1 | 18 - 19 | 2390 | 57 | |
XM_017018041.1 | XP_016873530.1 | 17 - 18 | 2448 | 57 | |
XM_017018047.1 | XP_016873536.1 | 17 - 18 | 2891 | 57 | |
XM_017018048.1 | XP_016873537.1 | 16 - 17 | 2847 | 57 | |
XM_017018051.1 | XP_016873540.1 | 14 - 15 | 1768 | 57 | |
GenBank mRNA | AB033027.1 | - | 16 - 17 | 2068 | 57 |
AK127457.1 | - | 17 - 18 | 2426 | 57 | |
BC105741.1 | - | 16 - 17 | 2219 | 57 | |
BC105931.1 | - | 16 - 17 | 2219 | 57 | |
BC105932.1 | - | 16 - 17 | 2219 | 57 | |
BC107480.1 | - | 16 - 17 | 2219 | 57 |