Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLAIN1
Entrez Gene ID
122060
Gene Name
SLAIN motif family member 1
Gene Aliases
C13orf32
Chromosome Location
Chr.13: 77697854 - 77764242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242868.1 NP_001229797.1 1 - 2 685 93
GenBank mRNA BG391962.1 - - 64 93
BQ433296.1 - 1 - 2 217 93

View Full Product Details