Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMT1L
Entrez Gene ID
81627
Gene Name
tRNA methyltransferase 1 like
Gene Aliases
C1orf25, MST070, MSTP070, TRM1L, bG120K12.3
Chromosome Location
Chr.1: 185118085 - 185157098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202423.1 NP_001189352.1 7 - 8 738 78
NM_030934.4 NP_112196.3 7 - 8 1130 78
XM_011510028.2 XP_011508330.1 7 - 8 2567 78
XM_017002419.1 XP_016857908.1 7 - 8 1102 78
GenBank mRNA AF288399.1 - 7 - 8 1052 78
AK074936.1 - 7 - 8 1079 78
AK074993.1 - 7 - 8 1102 78
AK098359.1 - 7 - 8 1097 78
AK131460.1 - 6 - 7 1772 78
AK226103.1 - 7 - 8 845 78
AK302166.1 - 8 - 9 662 78
AK312801.1 - 7 - 8 1099 78
BC035430.1 - 7 - 8 1082 78
BC039738.1 - 7 - 8 1093 78
BC045535.1 - 7 - 8 1079 78
BC053961.1 - 7 - 8 1079 78

View Full Product Details