The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007115.3 | NP_009046.2 | 5 - 6 | 81 | |
GenBank mRNA | AF086484.1 | - | 5 - 6 | 81 | |
AJ419936.1 | - | 5 - 6 | 81 | ||
AJ421518.1 | - | 5 - 6 | 81 | ||
BC030205.1 | - | 5 - 6 | 81 | ||
CA307541.1 | - | 1 - 2 | 700 | 81 | |
M31165.1 | - | 5 - 6 | 81 |