The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025176.5 | NP_079452.3 | 17 - 18 | 3334 | 151 |
XM_011529186.2 | XP_011527488.1 | 18 - 19 | 4173 | 151 | |
XM_011529187.2 | XP_011527489.1 | 18 - 19 | 4158 | 151 | |
XM_011529188.1 | XP_011527490.1 | 18 - 19 | 3550 | 151 | |
XM_011529189.1 | XP_011527491.1 | 18 - 19 | 3558 | 151 | |
XM_011529190.1 | XP_011527492.1 | 16 - 17 | 3272 | 151 | |
XM_011529191.1 | XP_011527493.1 | 14 - 15 | 3065 | 151 | |
XM_011529192.1 | XP_011527494.1 | 11 - 12 | 3985 | 151 | |
XM_011529194.2 | XP_011527496.1 | 9 - 10 | 3254 | 151 | |
XM_011529195.2 | XP_011527497.1 | 8 - 9 | 2079 | 151 | |
XM_011529197.2 | XP_011527499.1 | 10 - 11 | 2448 | 151 | |
XM_011529198.1 | XP_011527500.1 | 4 - 5 | 1620 | 151 | |
XM_017027732.1 | XP_016883221.1 | 19 - 20 | 5492 | 151 | |
XM_017027733.1 | XP_016883222.1 | 19 - 20 | 3604 | 151 | |
XM_017027734.1 | XP_016883223.1 | 11 - 12 | 2438 | 151 | |
XM_017027735.1 | XP_016883224.1 | 8 - 9 | 2084 | 151 | |
GenBank mRNA | AB023197.1 | - | 17 - 18 | 3320 | 151 |
BC036380.2 | - | 16 - 17 | 3266 | 151 |