The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039966.1 | NP_001035055.1 | 2 - 3 | 563 | 60 |
NM_001098201.1 | NP_001091671.1 | 1 - 2 | 236 | 60 | |
GenBank mRNA | AK289588.1 | - | 1 - 2 | 210 | 60 |
DC381205.1 | - | 1 - 2 | 238 | 60 | |
U63917.1 | - | 1 - 2 | 183 | 60 |