Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC28
Entrez Gene ID
23331
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
TPRBK
Chromosome Location
Chr.22: 27978014 - 28679865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145418.1 NP_001138890.1 12 - 13 4075 114
XM_005261405.2 XP_005261462.1 12 - 13 4075 114
XM_006724171.3 XP_006724234.1 11 - 12 12853 114
XM_011530018.2 XP_011528320.1 12 - 13 4254 114
XM_011530019.2 XP_011528321.1 12 - 13 4075 114
XM_011530020.1 XP_011528322.1 12 - 13 4075 114
XM_011530021.2 XP_011528323.1 12 - 13 4075 114
XM_011530022.1 XP_011528324.1 12 - 13 4075 114
XM_017028673.1 XP_016884162.1 12 - 13 4048 114
GenBank mRNA AB028966.3 - 7 - 8 2868 114
AB665284.1 - 12 - 13 4023 114
BC016465.1 - 4 - 5 619 114

View Full Product Details