Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
Chromosome Location
Chr.11: 26189123 - 26663289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313726.1 NP_001300655.1 13 - 14 1525 96
NM_001313727.1 NP_001300656.1 9 - 10 1083 96
NM_031418.3 NP_113606.2 12 - 13 1433 96
XM_017018118.1 XP_016873607.1 10 - 11 1061 96
XM_017018119.1 XP_016873608.1 7 - 8 673 96
GenBank mRNA AJ300461.1 - 12 - 13 2140 96
AK295143.1 - 9 - 10 996 96
AK295816.1 - 9 - 10 995 96
AK302659.1 - 12 - 13 1472 96
AK304781.1 - 13 - 14 1366 96
AK316018.1 - 9 - 10 995 96

View Full Product Details