The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300752.1 | NP_001287681.1 | 2 - 3 | 894 | 142 |
NM_001300759.1 | NP_001287688.1 | 3 - 4 | 913 | 142 | |
NM_018700.3 | NP_061170.2 | 3 - 4 | 1129 | 142 | |
XM_017009621.1 | XP_016865110.1 | 5 - 6 | 1141 | 142 | |
XM_017009622.1 | XP_016865111.1 | 5 - 6 | 1858 | 142 | |
XM_017009623.1 | XP_016865112.1 | 5 - 6 | 1141 | 142 | |
GenBank mRNA | AB196558.1 | - | 2 - 3 | 385 | 142 |
AJ272269.1 | - | 3 - 4 | 746 | 142 | |
AK296389.1 | - | 3 - 4 | 911 | 142 | |
AK316422.1 | - | 3 - 4 | 816 | 142 | |
BC130334.1 | - | 3 - 4 | 798 | 142 |