The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006108.3 | NP_006099.2 | 12 - 13 | 1927 | 62 |
GenBank mRNA | AB018305.2 | - | 11 - 12 | 1412 | 62 |
AK001584.1 | - | 4 - 5 | 496 | 62 | |
AK074803.1 | - | 12 - 13 | 1873 | 62 | |
AK291780.1 | - | 12 - 13 | 1875 | 62 | |
BC136513.1 | - | 12 - 13 | 1869 | 62 | |
BC136563.1 | - | 12 - 13 | 1869 | 62 |