Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRX1
Entrez Gene ID
79671
Gene Name
NLR family member X1
Gene Aliases
CLR11.3, DLNB26, NOD26, NOD5, NOD9
Chromosome Location
Chr.11: 119168334 - 119184016 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006718904.1 XP_006718967.1 9 - 10 3115 135
GenBank mRNA BC013199.1 - 5 - 6 1961 135

View Full Product Details