Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRX1
Entrez Gene ID
79671
Gene Name
NLR family member X1
Gene Aliases
CLR11.3, DLNB26, NOD26, NOD5, NOD9
Chromosome Location
Chr.11: 119168334 - 119184016 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282143.1 NP_001269072.1 9 - 10 2832 111
NM_001282144.1 NP_001269073.1 9 - 10 3240 111
NM_001282358.1 NP_001269287.1 9 - 10 2764 111
NM_024618.3 NP_078894.2 9 - 10 2859 111
XM_005271669.1 XP_005271726.1 9 - 10 3083 111
XM_005271670.1 XP_005271727.1 8 - 9 2736 111
XM_005271671.1 XP_005271728.1 8 - 9 2910 111
XM_005271672.1 XP_005271729.1 8 - 9 2733 111
XM_011542980.1 XP_011541282.1 9 - 10 2747 111
GenBank mRNA AB094095.1 - 9 - 10 2843 111
AK025131.1 - 4 - 5 1486 111
AK056454.1 - 8 - 9 2557 111
AK095247.1 - 7 - 8 2044 111
AK225223.1 - 4 - 5 1486 111
AK291716.1 - 9 - 10 2826 111
AY245437.1 - 8 - 9 2602 111
BC110890.1 - 9 - 10 3205 111
BX647705.1 - 9 - 10 2817 111
EF452237.1 - 8 - 9 2602 111

View Full Product Details