Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSG1
Entrez Gene ID
83445
Gene Name
germ cell associated 1
Gene Aliases
-
Chromosome Location
Chr.12: 13083537 - 13103720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080554.2 NP_001074023.1 6 - 7 1058 73
NM_001080555.2 NP_001074024.1 6 - 7 934 73
NM_001206842.1 NP_001193771.1 5 - 6 865 73
NM_001206843.1 NP_001193772.1 5 - 6 757 73
NM_001206845.1 NP_001193774.1 4 - 5 688 73
NM_031289.3 NP_112579.2 5 - 6 965 73
NM_153823.3 NP_722545.2 5 - 6 841 73
XM_005253493.2 XP_005253550.1 6 - 7 988 73
XM_005253495.1 XP_005253552.1 6 - 7 1112 73
XM_011520858.1 XP_011519160.1 6 - 7 1015 73
XM_011520859.1 XP_011519161.1 6 - 7 932 73
XM_011520860.1 XP_011519162.1 6 - 7 1139 73
XM_011520861.1 XP_011519163.1 5 - 6 916 73
XM_011520862.1 XP_011519164.1 6 - 7 1056 73
XM_011520863.1 XP_011519165.1 5 - 6 1040 73
GenBank mRNA AB593019.1 - 5 - 6 837 73
AB593020.1 - 5 - 6 713 73
AK027894.1 - 5 - 6 934 73
AK058045.1 - 6 - 7 1007 73
AK075288.1 - 5 - 6 835 73
AK075311.1 - 4 - 5 530 73
AK075322.1 - 5 - 6 711 73
AY358513.1 - 5 - 6 819 73
BC001796.1 - 5 - 6 911 73
BC033854.1 - 5 - 6 808 73
BC112896.1 - 5 - 6 625 73

View Full Product Details