The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135862.2 | NP_001129334.1 | 3 - 4 | 225 | 80 |
NM_001242450.1 | NP_001229379.1 | 2 - 3 | 265 | 80 | |
NM_138415.4 | NP_612424.1 | 2 - 3 | 270 | 80 | |
NR_134535.1 | 2 - 3 | 180 | 80 | ||
XM_006724122.1 | XP_006724185.1 | 2 - 3 | 405 | 80 | |
XM_011529855.1 | XP_011528157.1 | 2 - 3 | 405 | 80 | |
GenBank mRNA | AK092243.1 | - | 3 - 4 | 225 | 80 |
AK297317.1 | - | 2 - 3 | 258 | 80 | |
AK299836.1 | - | 2 - 3 | 221 | 80 | |
BC012187.1 | - | 2 - 3 | 270 | 80 | |
CR456448.1 | - | 2 - 3 | 152 | 80 | |
KU178842.1 | - | 2 - 3 | 119 | 80 | |
KU178843.1 | - | 2 - 3 | 119 | 80 | |
KU178844.1 | - | 2 - 3 | 119 | 80 |