Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINE1
Entrez Gene ID
5054
Gene Name
serpin family E member 1
Gene Aliases
PAI, PAI-1, PAI1, PLANH1
Chromosome Location
Chr.7: 101127087 - 101139266 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000602.4 NP_000593.1 4 - 5 864 75
XM_017012260.1 XP_016867749.1 4 - 5 866 75
GenBank mRNA AK293248.1 - 2 - 3 173 75
AK298048.1 - 4 - 5 667 75
AK316225.1 - 5 - 6 732 75
BC010860.1 - 4 - 5 829 75
CN309038.1 - 1 - 2 209 75
M14083.1 - 3 - 4 650 75
M16006.1 - 4 - 5 782 75
X04429.1 - 4 - 5 833 75
X04744.1 - 3 - 4 659 75
X12701.1 - 4 - 5 833 75

View Full Product Details