The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000602.4 | NP_000593.1 | 5 - 6 | 1054 | 58 |
XM_017012260.1 | XP_016867749.1 | 5 - 6 | 1056 | 58 | |
GenBank mRNA | AK293248.1 | - | 3 - 4 | 363 | 58 |
AK297728.1 | - | 6 - 7 | 996 | 58 | |
AK298048.1 | - | 5 - 6 | 857 | 58 | |
AK316225.1 | - | 6 - 7 | 922 | 58 | |
BC010860.1 | - | 5 - 6 | 1019 | 58 | |
CN309038.1 | - | 2 - 3 | 399 | 58 | |
M14083.1 | - | 4 - 5 | 840 | 58 | |
M16006.1 | - | 5 - 6 | 972 | 58 | |
X04429.1 | - | 5 - 6 | 1023 | 58 | |
X04744.1 | - | 4 - 5 | 849 | 58 | |
X12701.1 | - | 5 - 6 | 1023 | 58 |